Assay Details
| Gene Symbol: Socs16D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272733.2 | NP_001259662.1 | 2 - 3 | 587 | 82 | |
| NM_078666.4 | NP_523390.3 | 2 - 3 | 543 | 82 | ||
More Information
Set Membership: |
|

Socs16D
Genomic Map