Assay Details
| Gene Symbol: Socs16D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078666.4 | NP_523390.3 | 3 - 4 | 2647 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs16D| Gene Symbol: Socs16D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078666.4 | NP_523390.3 | 3 - 4 | 2647 | 53 | |
Set Membership: |
|