Assay Details
| Gene Symbol: Socs16D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272733.2 | NP_001259662.1 | 4 - 5 | 2822 | 88 | |
| NM_078666.4 | NP_523390.3 | 4 - 5 | 2763 | 88 | ||
More Information
Set Membership: |
|

Socs16D
Genomic Map