Assay Details
| Gene Symbol: wupA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258803.2 | NP_001245732.1 | 5 - 6 | 591 | 71 | |
| NM_078674.4 | NP_523398.1 | 4 - 5 | 408 | 71 | ||
| NM_167603.3 | NP_728137.1 | 3 - 4 | 377 | 71 | ||
| NM_167608.3 | NP_728142.1 | 5 - 6 | 591 | 71 | ||
| GenBank mRNA | BT125790.1 | - | 5 - 6 | 560 | 71 | |
| BT125851.1 | - | 4 - 5 | 381 | 71 | ||
| X59376.1 | - | 353 | 71 | |||
More Information
Set Membership: |
|

wupA
Genomic Map