Assay Details
| Gene Symbol: Cyp18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078679.3 | NP_523403.2 | 4 - 5 | 2032 | 65 | |
| NM_167628.2 | NP_728191.1 | 4 - 5 | 1734 | 65 | ||
More Information
Set Membership: |
|

Cyp18a1
Genomic Map