Assay Details
| Gene Symbol: CCKLR-17D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103551.2 | NP_001097021.1 | 3 - 4 | 966 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCKLR-17D1| Gene Symbol: CCKLR-17D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103551.2 | NP_001097021.1 | 3 - 4 | 966 | 58 | |
Set Membership: |
|