Assay Details
| Gene Symbol: pcm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169325.2 | NP_001162796.1 | 8 - 9 | 4203 | 165 | |
| NM_078684.4 | NP_523408.2 | 8 - 9 | 4203 | 165 | ||
| GenBank mRNA | AY051675.1 | - | 8 - 9 | 4143 | 165 | |
More Information
Set Membership: |
|

pcm
Genomic Map