Assay Details
| Gene Symbol: Grip84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169326.2 | NP_001162797.1 | 4 - 5 | 2433 | 72 | |
| NM_001169327.2 | NP_001162798.1 | 4 - 5 | 2433 | 72 | ||
| NM_001298515.1 | NP_001285444.1 | 3 - 4 | 2373 | 72 | ||
| NM_078685.3 | NP_523409.1 | 3 - 4 | 2373 | 72 | ||
| NM_167662.2 | NP_728264.1 | 4 - 5 | 2433 | 72 | ||
| NM_167663.2 | NP_728265.1 | 3 - 4 | 2373 | 72 | ||
| GenBank mRNA | AF118379.1 | - | 3 - 4 | 2197 | 72 | |
| BT150451.1 | - | 4 - 5 | 2266 | 72 | ||
More Information
Set Membership: |
|

Grip84
Genomic Map