Assay Details
| Gene Symbol: Zw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298527.1 | NP_001285456.1 | 3 - 4 | 551 | 70 | |
| NM_078687.2 | NP_523411.1 | 3 - 4 | 846 | 70 | ||
| NM_167676.2 | NP_728287.1 | 3 - 4 | 823 | 70 | ||
More Information
Set Membership: |
|

Zw
Genomic Map