Assay Details
| Gene Symbol: amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078692.2 | NP_523416.2 | 1 | 3891 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

amn| Gene Symbol: amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078692.2 | NP_523416.2 | 1 | 3891 | 146 | |
Set Membership: |
|