Assay Details
| Gene Symbol: shakB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144750.2 | NP_001138222.1 | 1 - 2 | 252 | 104 | |
| NM_001258858.2 | NP_001245787.1 | 5 - 6 | 1363 | 104 | ||
| NM_001258859.2 | NP_001245788.1 | 6 - 7 | 2227 | 104 | ||
| NM_001298574.1 | NP_001285503.1 | 5 - 6 | 2468 | 104 | ||
| NM_078701.3 | NP_523425.2 | 4 - 5 | 1659 | 104 | ||
| NM_134566.3 | NP_608410.2 | 3 - 4 | 763 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001144750.2 transcripts.
Set Membership: |
|

shakB
Genomic Map