Assay Details
| Gene Symbol: l(2)gl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258872.2 | NP_001245801.1 | 3 - 4 | 540 | 113 | |
| NM_001258873.2 | NP_001245802.1 | 2 - 3 | 404 | 113 | ||
| NM_001258874.2 | NP_001245803.1 | 3 - 4 | 615 | 113 | ||
| NM_001258875.2 | NP_001245804.1 | 4 - 5 | 753 | 113 | ||
| NM_001272858.1 | NP_001259787.1 | 3 - 4 | 615 | 113 | ||
| NM_078715.4 | NP_523439.2 | 2 - 3 | 473 | 113 | ||
| NM_164349.3 | NP_722584.1 | 3 - 4 | 616 | 113 | ||
| GenBank mRNA | AY051654.1 | - | 2 - 3 | 463 | 113 | |
More Information
Set Membership: |
|

l(2)gl
Genomic Map