Assay Details
| Gene Symbol: l(2)gl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258872.2 | NP_001245801.1 | 7 - 8 | 3262 | 94 | |
| NM_001258873.2 | NP_001245802.1 | 6 - 7 | 3126 | 94 | ||
| NM_001258874.2 | NP_001245803.1 | 7 - 8 | 3337 | 94 | ||
| NM_001258875.2 | NP_001245804.1 | 8 - 9 | 3475 | 94 | ||
| NM_001272858.1 | NP_001259787.1 | 7 - 8 | 3337 | 94 | ||
| NM_078715.4 | NP_523439.2 | 6 - 7 | 3195 | 94 | ||
| NM_164348.3 | NP_722583.1 | 6 - 7 | 3102 | 94 | ||
| NM_164349.3 | NP_722584.1 | 7 - 8 | 3338 | 94 | ||
| NM_164350.3 | NP_722585.1 | 6 - 7 | 3177 | 94 | ||
| NM_164351.3 | NP_722586.1 | 6 - 7 | 3172 | 94 | ||
| NM_164352.3 | NP_722587.1 | 7 - 8 | 3029 | 94 | ||
| GenBank mRNA | AY051654.1 | - | 6 - 7 | 3185 | 94 | |
| BT100129.1 | - | 6 - 7 | 3098 | 94 | ||
More Information
Set Membership: |
|

l(2)gl
Genomic Map