Assay Details
| Gene Symbol: l(2)gl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258872.2 | NP_001245801.1 | 9 - 10 | 3813 | 167 | |
| NM_001258873.2 | NP_001245802.1 | 8 - 9 | 3677 | 167 | ||
| NM_001258874.2 | NP_001245803.1 | 9 - 10 | 3888 | 167 | ||
| NM_001258875.2 | NP_001245804.1 | 10 - 11 | 4026 | 167 | ||
| NM_078715.4 | NP_523439.2 | 8 - 9 | 3746 | 167 | ||
| NM_164348.3 | NP_722583.1 | 8 - 9 | 3653 | 167 | ||
| NM_164349.3 | NP_722584.1 | 9 - 10 | 3889 | 167 | ||
| NM_164350.3 | NP_722585.1 | 8 - 9 | 3728 | 167 | ||
| NM_164351.3 | NP_722586.1 | 8 - 9 | 3723 | 167 | ||
| NM_164352.3 | NP_722587.1 | 9 - 10 | 3580 | 167 | ||
| GenBank mRNA | AY051654.1 | - | 8 - 9 | 3736 | 167 | |
| BT100129.1 | - | 8 - 9 | 3649 | 167 | ||
More Information
Set Membership: |
|

l(2)gl
Genomic Map