Assay Details
| Gene Symbol: kis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258889.1 | NP_001245818.1 | 11 - 12 | 9668 | 116 | |
| NM_001258890.1 | NP_001245819.1 | 9 - 10 | 9395 | 116 | ||
| NM_001258891.2 | NP_001245820.1 | 10 - 11 | 9786 | 116 | ||
| NM_078717.3 | NP_523441.1 | 10 - 11 | 9605 | 116 | ||
| GenBank mRNA | AF215703.1 | - | 10 - 11 | 9577 | 116 | |
| MN044634.1 | - | 10 - 11 | 9650 | 116 | ||
More Information
Set Membership: |
|

kis
Genomic Map