Assay Details
| Gene Symbol: kis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144289.2 | NP_001137761.1 | 14 - 15 | 13435 | 104 | |
| NM_001258889.1 | NP_001245818.1 | 16 - 17 | 12913 | 104 | ||
| NM_001258890.1 | NP_001245819.1 | 14 - 15 | 12640 | 104 | ||
| NM_001258891.2 | NP_001245820.1 | 14 - 15 | 12638 | 104 | ||
| NM_078717.3 | NP_523441.1 | 15 - 16 | 12850 | 104 | ||
| NM_164376.2 | NP_722617.1 | 4 - 5 | 3132 | 104 | ||
| GenBank mRNA | AF215703.1 | - | 15 - 16 | 12822 | 104 | |
| MN044634.1 | - | 15 - 16 | 12895 | 104 | ||
More Information
Set Membership: |
|

kis
Genomic Map