Assay Details
| Gene Symbol: ds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298622.1 | NP_001285551.1 | 4 - 5 | 3971 | 120 | |
| NM_078722.3 | NP_523446.2 | 4 - 5 | 3971 | 120 | ||
More Information
Set Membership: |
|

ds
Genomic Map