Assay Details
| Gene Symbol: aop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272979.1 | NP_001259908.1 | 3 - 4 | 2243 | 102 | |
| NM_001272980.1 | NP_001259909.1 | 2 - 3 | 1796 | 102 | ||
| NM_001272981.1 | NP_001259910.1 | 2 - 3 | 2186 | 102 | ||
| NM_078731.3 | NP_523455.2 | 2 - 3 | 2186 | 102 | ||
| NM_164457.2 | NP_722766.1 | 2 - 3 | 1573 | 102 | ||
| GenBank mRNA | BT015272.1 | - | 2 - 3 | 2133 | 102 | |
More Information
Set Membership: |
|

aop
Genomic Map