Assay Details
| Gene Symbol: lilli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144300.2 | NP_001137772.1 | 8 - 9 | 4659 | 63 | |
| NM_001144301.1 | NP_001137773.1 | 9 - 10 | 4999 | 63 | ||
| NM_001144302.2 | NP_001137774.1 | 8 - 9 | 4768 | 63 | ||
| NM_001144303.1 | NP_001137775.1 | 10 - 11 | 6303 | 63 | ||
| NM_001273044.1 | NP_001259973.1 | 9 - 10 | 6456 | 63 | ||
| NM_001347818.1 | NP_001334729.1 | 9 - 10 | 6456 | 63 | ||
| NM_078740.3 | NP_523464.1 | 10 - 11 | 7219 | 63 | ||
| NM_164516.2 | NP_722863.1 | 11 - 12 | 7387 | 63 | ||
| NM_164517.1 | NP_722864.1 | 10 - 11 | 6108 | 63 | ||
| GenBank mRNA | AF289034.1 | - | 11 - 12 | 7364 | 63 | |
| AF293971.1 | - | 10 - 11 | 7193 | 63 | ||
More Information
Set Membership: |
|

lilli
Genomic Map