Assay Details
| Gene Symbol: gkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078741.4 | NP_523465.2 | 2 - 3 | 747 | 63 | |
| GenBank mRNA | AY051884.1 | - | 2 - 3 | 714 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gkt| Gene Symbol: gkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078741.4 | NP_523465.2 | 2 - 3 | 747 | 63 | |
| GenBank mRNA | AY051884.1 | - | 2 - 3 | 714 | 63 | |
Set Membership: |
|