Assay Details
| Gene Symbol: Ptpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078742.3 | NP_523466.2 | 2 - 3 | 745 | 72 | |
| GenBank mRNA | AY060614.1 | - | 2 - 3 | 739 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpa| Gene Symbol: Ptpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078742.3 | NP_523466.2 | 2 - 3 | 745 | 72 | |
| GenBank mRNA | AY060614.1 | - | 2 - 3 | 739 | 72 | |
Set Membership: |
|