Assay Details
| Gene Symbol: GluRIIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078760.3 | NP_523484.2 | 7 - 8 | 1559 | 66 | |
| GenBank mRNA | BT010006.1 | - | 7 - 8 | 1557 | 66 | |
More Information
Set Membership: |
|

GluRIIA
Genomic Map