Assay Details
| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014477.2 | NP_001014477.1 | 2 - 3 | 616 | 57 | |
| NM_001298769.1 | NP_001285698.1 | 2 - 3 | 616 | 57 | ||
| NM_078773.3 | NP_523497.2 | 2 - 3 | 616 | 57 | ||
| GenBank mRNA | BT030993.1 | - | 2 - 3 | 427 | 57 | |
| BT031204.1 | - | 2 - 3 | 596 | 57 | ||
More Information
Set Membership: |
|

Gart
Genomic Map