Assay Details
| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014477.2 | NP_001014477.1 | 3 - 4 | 1267 | 61 | |
| NM_001298769.1 | NP_001285698.1 | 3 - 4 | 1267 | 61 | ||
| NM_078773.3 | NP_523497.2 | 3 - 4 | 1267 | 61 | ||
| GenBank mRNA | BT030993.1 | - | 3 - 4 | 1078 | 61 | |
| BT031204.1 | - | 3 - 4 | 1247 | 61 | ||
More Information
Set Membership: |
|

Gart
Genomic Map