Assay Details
| Gene Symbol: ninaC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273260.1 | NP_001260189.1 | 12 - 13 | 4479 | 97 | |
| NM_078779.3 | NP_523503.2 | 12 - 13 | 4027 | 97 | ||
More Information
Set Membership: |
|

ninaC
Genomic Map