Assay Details
| Gene Symbol: L2HGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299147.1 | NP_001286076.1 | 1 - 2 | 267 | 72 | |
| NM_001299148.1 | NP_001286077.1 | 1 - 2 | 452 | 72 | ||
| NM_136079.3 | NP_609923.2 | 1 - 2 | 527 | 72 | ||
| GenBank mRNA | BT044190.1 | - | 1 - 2 | 378 | 72 | |
More Information
Set Membership: |
|

L2HGDH
Genomic Map