Assay Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136090.3 | NP_609934.3 | 1 - 2 | 775 | 97 | |
| GenBank mRNA | AY071630.1 | - | 1 - 2 | 600 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc19| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136090.3 | NP_609934.3 | 1 - 2 | 775 | 97 | |
| GenBank mRNA | AY071630.1 | - | 1 - 2 | 600 | 97 | |
Set Membership: |
|