Assay Details
| Gene Symbol: tj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273653.1 | NP_001260582.1 | 1 | 1960 | 94 | |
| NM_136125.3 | NP_609969.2 | 1 | 1960 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tj| Gene Symbol: tj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273653.1 | NP_001260582.1 | 1 | 1960 | 94 | |
| NM_136125.3 | NP_609969.2 | 1 | 1960 | 94 | ||
Set Membership: |
|