Assay Details
| Gene Symbol: Pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273690.1 | NP_001260619.1 | 2 | 1479 | 73 | |
| NM_136179.3 | NP_610023.1 | 1 | 1448 | 73 | ||
| GenBank mRNA | BT022436.1 | - | 1 | 1399 | 73 | |
More Information
Set Membership: |
|

Pyroxd1
Genomic Map