Assay Details
| Gene Symbol: twit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136223.3 | NP_610067.1 | 2 - 3 | 453 | 58 | |
| GenBank mRNA | AY118565.1 | - | 2 - 3 | 423 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

twit| Gene Symbol: twit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136223.3 | NP_610067.1 | 2 - 3 | 453 | 58 | |
| GenBank mRNA | AY118565.1 | - | 2 - 3 | 423 | 58 | |
Set Membership: |
|