Assay Details
| Gene Symbol: Mtp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136231.4 | NP_610075.2 | 2 - 3 | 988 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtp| Gene Symbol: Mtp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136231.4 | NP_610075.2 | 2 - 3 | 988 | 71 | |
Set Membership: |
|