Assay Details
| Gene Symbol: Mtp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136231.4 | NP_610075.2 | 4 - 5 | 2459 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtp| Gene Symbol: Mtp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136231.4 | NP_610075.2 | 4 - 5 | 2459 | 74 | |
Set Membership: |
|