Assay Details
| Gene Symbol: CG8677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299194.1 | NP_001286123.1 | 2 - 3 | 655 | 98 | |
| NM_136257.4 | NP_610101.3 | 2 - 3 | 655 | 98 | ||
More Information
Set Membership: |
|

CG8677
Genomic Map