Assay Details
| Gene Symbol: Clamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014498.3 | NP_001014498.1 | 3 - 4 | 717 | 105 | |
| NM_136293.4 | NP_610137.1 | 3 - 4 | 717 | 105 | ||
| GenBank mRNA | BT001676.1 | - | 3 - 4 | 716 | 105 | |
| BT001891.1 | - | 1932 | 105 | |||
More Information
Set Membership: |
|

Clamp
Genomic Map