Assay Details
| Gene Symbol: Clamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014498.3 | NP_001014498.1 | 4 - 5 | 1154 | 75 | |
| NM_136293.4 | NP_610137.1 | 4 - 5 | 1154 | 75 | ||
| GenBank mRNA | BT001676.1 | - | 4 - 5 | 1153 | 75 | |
| BT001891.1 | - | 2369 | 75 | |||
More Information
Set Membership: |
|

Clamp
Genomic Map