Assay Details
| Gene Symbol: Clamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136293.4 | NP_610137.1 | 5 - 6 | 1419 | 89 | |
| GenBank mRNA | BT001891.1 | - | 2634 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clamp| Gene Symbol: Clamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136293.4 | NP_610137.1 | 5 - 6 | 1419 | 89 | |
| GenBank mRNA | BT001891.1 | - | 2634 | 89 | ||
Set Membership: |
|