Assay Details
| Gene Symbol: TpnC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169584.2 | NP_001163055.1 | 3 - 4 | 117 | 66 | |
| NM_136329.3 | NP_610173.2 | 2 - 3 | 220 | 66 | ||
| GenBank mRNA | AJ512938.1 | - | 2 - 3 | 206 | 66 | |
| AY283602.1 | - | 3 - 4 | 114 | 66 | ||
| BT044320.1 | - | 2 - 3 | 231 | 66 | ||
| KX531829.1 | - | 1 - 2 | 40 | 66 | ||
More Information
Set Membership: |
|

TpnC4
Genomic Map