Assay Details
| Gene Symbol: Not3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273783.1 | NP_001260712.1 | 3 - 4 | 858 | 71 | |
| NM_136332.3 | NP_610176.1 | 3 - 4 | 858 | 71 | ||
| GenBank mRNA | AY069428.1 | - | 3 - 4 | 836 | 71 | |
| BT083390.1 | - | 3 - 4 | 818 | 71 | ||
More Information
Set Membership: |
|

Not3
Genomic Map