Assay Details
| Gene Symbol: l(2)09851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273784.1 | NP_001260713.1 | 1 | 1473 | 101 | |
| NM_136338.3 | NP_610182.3 | 1 | 1473 | 101 | ||
| GenBank mRNA | AY071477.1 | - | 1 | 1457 | 101 | |
| AY128462.1 | - | 1 | 1457 | 101 | ||
More Information
Set Membership: |
|

l(2)09851
Genomic Map