Assay Details
| Gene Symbol: Ptr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259216.2 | NP_001246145.1 | 3 - 4 | 892 | 68 | |
| NM_001259217.2 | NP_001246146.1 | 3 - 4 | 948 | 68 | ||
| NM_136365.3 | NP_610209.2 | 3 - 4 | 637 | 68 | ||
| GenBank mRNA | BT003500.1 | - | 3 - 4 | 635 | 68 | |
More Information
Set Membership: |
|

Ptr
Genomic Map