Assay Details
| Gene Symbol: Ptr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259216.2 | NP_001246145.1 | 4 - 5 | 1108 | 110 | |
| NM_001259217.2 | NP_001246146.1 | 4 - 5 | 1164 | 110 | ||
| NM_136365.3 | NP_610209.2 | 4 - 5 | 853 | 110 | ||
| GenBank mRNA | BT003500.1 | - | 4 - 5 | 851 | 110 | |
More Information
Set Membership: |
|

Ptr
Genomic Map