Assay Details
| Gene Symbol: Fmo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136373.4 | NP_610217.1 | 4 - 5 | 661 | 57 | |
| GenBank mRNA | AY058349.1 | - | 4 - 5 | 643 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmo-2| Gene Symbol: Fmo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136373.4 | NP_610217.1 | 4 - 5 | 661 | 57 | |
| GenBank mRNA | AY058349.1 | - | 4 - 5 | 643 | 57 | |
Set Membership: |
|