Assay Details
| Gene Symbol: Fmo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136373.4 | NP_610217.1 | 5 - 6 | 1194 | 69 | |
| GenBank mRNA | AY058349.1 | - | 5 - 6 | 1176 | 69 | |
More Information
Set Membership: |
|

Fmo-2
Genomic Map