Assay Details
| Gene Symbol: CG9447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136399.4 | NP_610243.4 | 4 - 5 | 1270 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG9447| Gene Symbol: CG9447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136399.4 | NP_610243.4 | 4 - 5 | 1270 | 66 | |
Set Membership: |
|