Assay Details
| Gene Symbol: Hsepi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169591.2 | NP_001163062.1 | 2 - 3 | 1802 | 100 | |
| NM_136405.4 | NP_610249.2 | 2 - 3 | 1808 | 100 | ||
| GenBank mRNA | BT082091.1 | - | 2 - 3 | 1767 | 100 | |
More Information
Set Membership: |
|

Hsepi
Genomic Map