Assay Details
| Gene Symbol: mEFTu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136444.3 | NP_610288.1 | 2 - 3 | 1423 | 72 | |
| GenBank mRNA | AY058330.1 | - | 2 - 3 | 1361 | 72 | |
| BT010319.1 | - | 2 - 3 | 1369 | 72 | ||
More Information
Set Membership: |
|

mEFTu2
Genomic Map