Assay Details
| Gene Symbol: PIG-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136448.3 | NP_610292.2 | 1 - 2 | 262 | 69 | |
| GenBank mRNA | BT149886.1 | - | 1 - 2 | 254 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIG-G| Gene Symbol: PIG-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136448.3 | NP_610292.2 | 1 - 2 | 262 | 69 | |
| GenBank mRNA | BT149886.1 | - | 1 - 2 | 254 | 69 | |
Set Membership: |
|