Assay Details
| Gene Symbol: PIG-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136448.3 | NP_610292.2 | 6 - 7 | 2110 | 64 | |
| GenBank mRNA | BT149886.1 | - | 6 - 7 | 2102 | 64 | |
More Information
Set Membership: |
|

PIG-G
Genomic Map