Assay Details
| Gene Symbol: azot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136492.4 | NP_610336.1 | 1 | 377 | 64 | |
| GenBank mRNA | AY070782.1 | - | 1 | 377 | 64 | |
| KX531992.1 | - | 1 | 327 | 64 | ||
More Information
Set Membership: |
|

azot
Genomic Map