Assay Details
| Gene Symbol: Vha44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144207.2 | NP_001137679.1 | 8 - 9 | 2336 | 61 | |
| NM_057918.5 | NP_477266.1 | 6 - 7 | 992 | 61 | ||
| NM_134313.4 | NP_599140.1 | 6 - 7 | 1047 | 61 | ||
| NM_166163.3 | NP_725564.1 | 7 - 8 | 1209 | 61 | ||
| NM_166164.5 | NP_725565.3 | 8 - 9 | 2391 | 61 | ||
| GenBank mRNA | AY061038.1 | - | 6 - 7 | 1026 | 61 | |
| AY102676.1 | - | 6 - 7 | 992 | 61 | ||
More Information
Set Membership: |
|

Vha44
Genomic Map